Photochemical Borosilicate Glass Immersion Well Without Joint, Ace Glass Incorporated
Supplier: Ace Glass
7875-35 7875-30 7875-45 7875-40
89061-382EA 466.45 USD InStock
89061-382 89061-384 89061-386 89061-388
Photochemical Borosilicate Glass Immersion Well Without Joint, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Double-walled, quartz, immersion well for use with reactors and lamps only
The well offers a greaseless connection and can be adjusted to desired height. Comes with or without #7 Ace Thred bushing. The Ace-Thred means easier dismantling since there is no chance of freezing.
A small diameter inner tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well is used with 89059-230 nylon bushing with FETFE O-Ring to form a compression type seal with threaded reaction vessels.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty