Human Unprocessed Bone Marrow, Charles River Laboratories Cell Solutions
Supplier: CHARLES RIVER LABS CELL SOLUTION MS
BM008F-3 BM008F-4
101222-912EA 4684.62 USD
101222-912 101222-914
Human Unprocessed Bone Marrow, Charles River Laboratories Cell Solutions
Cells
Human unprocessed bone marrow is collected via our qualified hospital and physician network from healthy human volunteer donors
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
Bone marrow aspirates and cells are drawn from the posterior iliac crest using a syringe containing anticoagulant. The donor is required to review and sign an approved procedure-specific consent form prior to the collection.