Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-COX2 Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
RP111 RP111-05
99991-388EA 849.01 USD InStock
99991-388 95024-978
Anti-COX2 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 70 kDa protein. COX-2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters and cytokines that induce its expression. COX-2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX-1 levels remained unchanged.

Type:
Antigen: COX2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty