Dumont Tweezers Style 11, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0305-11NI-CO 0305-11NI-PO
102091-728EA 46.62 USD InStock
102091-728 102091-730
Dumont Tweezers Style 11, Electron Microscopy Sciences
Forceps
Strong tips. Ideal to manipulate delicate pieces. Length: 120 mm.
Nickel is a 100% antimagnetic alloy composed of Cu, Ni, Mn and Zn. lt is widely appreciated as it does not damage the components handled by it.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty