Vikan® Tank Brushes, Hard, Remco Products
Supplier: Remco Products
70376 70372 70373 70374 70375 70379
76007-780EA 56.88 USD
76007-780 76007-932 76007-934 76007-936 76007-938 76007-940
Vikan® Tank Brushes, Hard, Remco Products
Brushes
The oval-shaped brush with stiff bristles is effective for scrubbing inside large industrial tanks and other large container-like equipment.
  • Block material is polypropylene; bristle material is polyester
  • Dishwasher safe
  • Autoclavable up to 250°F
The rounded edges work well to clean down the sides of equipment. It also fits well into large channel drains.
Información para pedidos: FDA compliant raw material.