Vikan® Tube Brushes, Remco Products
Supplier: Remco Products
53815 53825 53835 53865 53875 53885
76007-520EA 43.66 USD
76007-520 76007-522 76007-524 76007-526 76007-528 76007-530
Vikan® Tube Brushes, Remco Products
Brushes
This tube and valve brush is designed to clean hard to reach areas.
  • Dishwasher safe
  • Autoclavable up to 250 °F
Polypropylene block material; polyester bristle material. White color.
The bristles are attached to a helically wound stainless steel shaft, making it strong and chemically resistant. 
Certificaciones: FDA compliant raw material.