Urea ≥99.5%, Ultrapure for molecular biology
Supplier: THERMO SCIENTIFIC CHEMICALS INC MS
J75826-A7
J75826-A1
101173-314EA
474.78
USD
101173-314
101173-312
Urea ≥99.5%, Ultrapure for molecular biology
Urea
Formula:
NH₂CONH₂ MW: 60.06 g/mol Boiling Pt: 197 Melting Pt: 132.7 Density: 1.335 Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 14,9867 |
|
Specification Test Results
Assay Cyanate | None Detected |
Assay | ≥99.5% |
Chloride Content | ≤0.0005% |
Conductivity | ≤15 µmhos/cm |
DNase | Passes Test |
Identification (FTIR) | Conforms |
Residue on Ignition | ≤0.01% |
Impurity Content Sulfate (SO4) | ≤0.001% |
Insoluble Material | ≤0.01% |
Protease Activity | Passes Test |
RNase | Passes Test |
Spectral Ratio (260 nm, 6 M, 1 cm) | ≤0.055 |
Spectral Ratio (280 nm, 6 M, 1 cm) | ≤0.044 |
Copper (Cu) | ≤0.5 ppm |
Iron (Fe) | ≤0.5 ppm |
Lead (Pb) | ≤0.5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...