H62384-03
H62384-MD
AAH62384-03EA
120
USD
InStock
AAH62384-03
AAH62384-MD
Cis-4-Hydroxy-L-proline methyl ester hydrochloride 95%
cis-4-Hydroxy-L-proline methyl ester hydrochloride
cis-4-Hydroxy-L-proline methyl ester hydrochloride, 95%
Purity: 95%
CAS No.: 40126-30-5
Molecular Formula: C6H11NO3.HCl
Formula Weight: 181.62
MDL No.: MFCD00237835