Cefuroxime sodium salt 50 mg/ml distilled water, Filter sterile

Supplier: Origen Biomedical, Inc
Danger

J67116-8EQ J67116-XF
AAJ67116-8EQEA 182 USD
AAJ67116-8EQ AAJ67116-XF
Cefuroxime sodium salt 50 mg/ml distilled water, Filter sterile
Cefuroxime sodium salt

Cefuroxime sodium salt, 50 mg/ml in distilled water, sterile-filtered


CAS No.: 56238-63-2 Storage Temperature: -30°C to -10°C Physical Form: Liquid MDL No.: MFCD09878727

Formula: C₁₆H₁₅N₄NaO₈S
MW: 446.366 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878727
CAS Number: 56238-63-2
EINECS: 260-073-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR