(±)-2-Aminobutyric acid, powder

Supplier: E. Becker & Co. GmbH

0210043025 0210043080 0210043090
IC10043025EA 75.68 USD
IC10043025 IC10043080 IC10043090
(±)-2-Aminobutyric acid, powder
(±)-2-Aminobutyric acid

Soluble in water (5%). Sparingly soluble in alcohol; insoluble in ether.

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00008093
CAS Number: 2835-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR