Naphthol AS-D chloroacetate 98, powder

Supplier: E. Becker & Co. GmbH

02102424.1 02102424.5 0210242401
IC102424.1EA 74.34 USD
IC102424.1 IC102424.5 IC10242401
Naphthol AS-D chloroacetate 98, powder
Naphthol AS-D chloroacetate

Naphthol AS-D chloroacetate is a substrate for histochemical localization of esterases.


  • Presentation: White Powder


Soluble in ethyl acetate (5 mg/mL - clear, slightly yellow solution); methanol (1 mg/mL-clear, colorless solution) and acetone.


Naphthol AS-D chloroacetate is used as a substrate for esterases.

Formula: C₂₀H₁₆ClNO₃
MW: 353.8 g/mol
Melting Pt: 161…171 °C
Storage Temperature: Freezer
CAS Number: 35245-26-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a777a327ee2447a12204208cf626a4a16af2234&vid=0a777a327ee2447a12204208cf626a4a16af2234&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 161 - 171°C
UV/Vis: l max (methanol) 232 ± 5 nm
Free Naphthol ≤100 ppm
Purity (TLC) >98%
Solubility (5% in acetone) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR