Gum tragacanth, powder

Supplier: E. Becker & Co. GmbH

0210479280 0210479290
IC10479280EA 101.64 USD
IC10479280 IC10479290 IC10479291
Gum tragacanth, powder
Gum tragacanth

Tragacanth is used in pharmaceuticals and foods as an emulsifier, thickener, stabiliser, and texturant additive.


Insoluble in alcohol.


Tragacanth is used in pharmaceuticals and foods as an emulsifier, thickener, stabilizer, and texturant additive.


Precaución: Research Use Only (RUO).

MDL Number: MFCD00131255
CAS Number: 9000-65-1

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR