L-Aspartic acid magnesium salt, powder

Supplier: E. Becker & Co. GmbH

0215040625 0215040691
IC15040625EA 83.06 USD
IC15040625 IC15040691
L-Aspartic acid magnesium salt, powder
L-Aspartic acid magnesium salt

Freely soluble in water.

Formula: C₈H₁₂MgN₂O₈
MW: 288.495 g/mol
CAS Number: 18962-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR