Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hexacyanoferrate(III) ≥99%, dark red-orange crystalline powder ACS

Supplier: E. Becker & Co. GmbH

0215255980 0215255990
IC15255980EA 89.45 USD InStock
IC15255980 IC15255990
Potassium hexacyanoferrate(III) ≥99%, dark red-orange crystalline powder ACS
Potassium hexacyanoferrate(III)
Potassium ferricyanide is the chemical compound with the formula K3[Fe(CN)6]. This bright red salt contains the octahedrally coordinated [Fe(CN)6]3- ion. It is soluble in water and its solution shows some green-yellow fluorescence.

  • Auto Ignition: > 100 °C (Lit.)
  • Vapor Pressure: ca. .000001 hPa at 25 °C (Lit.)

Potassium ferricyanide is used as an oxidizing agent to remove silver from negatives and positives, a process called dot etching.

Solubility (10% soln): Yellow with no trace of green or brown

Potassium ferricyanide is used as an oxidizing agent to remove silver from negatives and positives, a process called dot etching.

Precaución: Research Use Only (RUO).


Formula: K₃Fe(CN)₆
Boiling Pt: 2735 °C (1013 hPa) (lit.)
Melting Pt: 1539 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Insoluble Matter ≤0.005%
Chloride (Cl) ≤0.01%
Sulfates (SO42-) ≤0.01%
Ferro Compounds ≤0.05%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty