Potassium iodide ≥99.0%, white solid ACS

Supplier: E. Becker & Co. GmbH
Warning

0215256280 0215256290
IC15256280EA 130.97 USD
IC15256280 76102-744
Potassium iodide ≥99.0%, white solid ACS
Potassium iodide

Potassium Iodide is known as Potassium Salt of Hydriodic Acid [HI]. Its Ionic form is K+ I-, and iodide ion being mild reducing agent, it can be easily oxidized to Iodine by powerful oxidizing agent like chlorine.


Potassium iodide is often used as a fluorescence quencher. The use of KI to facilitate the sorption of microbial proteins into dextran hydrogels from cell lysates has been described. KI has been applied in the elucidation of the crystal structure of the S100-A3 apoprotein. The use of KI in the isolation of single stranded DNA has been described.


Chloride & Bromide ≤0.01% ,Iodate (IO3) ≤3 ppm, Phosphate (PO4) ≤0.001% , Sulfate (SO4) ≤0.005%, Iron (Fe) ≤3 ppm ,Calcium (Ca) ≤0.002% ,Magnesium (Mg) ≤0.001% ,Sodium (Na) ≤0.005% ,Barium (Ba) ≤0.002%


Potassium iodide is often used as a fluorescence quencher. The use of KI to facilitate the sorption of microbial proteins into dextran hydrogels from cell lysates has been described. KI has been applied in the elucidation of the crystal structure of the S100-A3 apoprotein. The use of KI in the isolation of single stranded DNA has been described.

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686 (lit.)
Density: 1.32 g/mL (25 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
Merck Index: 13,07727