5-(N-2-3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis(2,3-dihydroxypropyl)isophthalamide, white powder
Supplier: E. Becker & Co. GmbH
Non-ionic monomeric triiodinated x-ray contrast agent
5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide is useful as a nonionic density gradient medium.
Density gradient medium to measure hemodynamic parameters in vivo. Induces apoptosis. Orally active.
Soluble in water (50 mg/ml). It is also soluble in formamide and dimethyformamide.
5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide is useful as a nonionic density gradient medium.
Formula:
C₁₉H₂₆I₃N₃O₉ MW: 821.142 g/mol |
MDL Number:
MFCD00077732 CAS Number: 66108-95-0 |
|
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Reference |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...