Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyanothiazole 95%

Supplier: Origen Biomedical, Inc
Warning

H63425-03 H63425-06 H63425-MD
AAH63425-03EA 173 USD InStock
AAH63425-03 AAH63425-06 AAH63425-MD
2-Cyanothiazole 95%
2-Cyanothiazole
2-Cyanothiazole, 95%

Purity: 95% CAS No.: 1452-16-0 Molecular Formula: C4H2N2S Formula Weight: 110.14 Physical Form: Solid UN No.: 3439 MDL No.: MFCD09702472


Formula: C4H2N2S
Storage Temperature: Ambient
MDL Number: MFCD09702472
CAS Number: 1452-16-0
UN: 3439
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty