Iridium(IV) chloride ≥99.95%

Supplier: BEANTOWN CHEMICAL - BTC

126325-1G 126325-250MG 126325-5G
BT126325-1GEA 360 USD
BT126325-1G BT126325-250MG BT126325-5G
Iridium(IV) chloride ≥99.95%
Iridium(IV) chloride

CAS: 10025-97-5; EC No: 233-048-8; MDL No: MFCD00016155; RTECS: NO3610000 Pieces/Powder; Molecular Formula: IrCl4; MW: 334.01 Melting Point: 350° Hygroscopic


  • RTECS: NO3610000
  • Hygroscopic
  • Pieces/Powder

Formula: Cl₄Ir
MW: 334.03
Melting Pt: >350 °C
MDL Number: MFCD00016155
CAS Number: 10025-97-5
EINECS: 233-048-8

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR