Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alizarin complexone, (max. 10% H₂O)

Supplier: BEANTOWN CHEMICAL - BTC

130475-1G 130475-5G
BT130475-1GEA 80.25 USD InStock
BT130475-1G BT130475-5G
Alizarin complexone, (max. 10% H₂O)
Alizarin complexone
CAS: 3952-78-1; EC No: 223-544-2; MDL No: MFCD00001202; RTECS: AH0585000 Powder; Molecular Formula: C19H15NO8; MW: 385.32

  • RTECS: AH0585000
  • Powder


Formula: C₁₉H₁₅NO₈
MDL Number: MFCD00001202
CAS Number: 3952-78-1
EINECS: 223-544-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty