Platinum ≥99.95% (trace metals basis), wire Ø 1.0 mm, annealed
Supplier: BEANTOWN CHEMICAL - BTC
130205-25CM
130205-5CM
BT130205-25CMEA
930
USD
BT130205-25CM
BT130205-5CM
Platinum ≥99.95% (trace metals basis), wire Ø 1.0 mm, annealed
Platinum
CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000
Solid; Molecular Formula: Pt; MW: 195.08
Melting Point: 1772°; Boiling Point: 3827°
Density (g/mL): 21.45
- RTECS: TP2160000
- Solid
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Density: 21.45 g/mL |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...