Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzophenone ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

134130-1KG 134130-250G 134130-50G
BT134130-1KGEA 666 USD InStock
BT134130-1KG BT134130-250G BT134130-50G
4-Hydroxybenzophenone ≥98%
4-Hydroxybenzophenone
CAS: 1137-42-4; EC No: 214-507-1; MDL No: MFCD00002355; RTECS: PC4959775 Powder; Molecular Formula: C13H10O2; MW: 198.22 Melting Point: 132-135°

  • RTECS: PC4959775
  • Powder


Formula: C₁₃H₁₀O₂
Melting Pt: 132…135 °C
MDL Number: MFCD00002355
CAS Number: 1137-42-4
EINECS: 214-507-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty