Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allantoin ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

134260-1KG 134260-250G 134260-5KG
BT134260-1KGEA 125 USD InStock
BT134260-1KG BT134260-250G BT134260-5KG
Allantoin ≥98%
Allantoin
CAS: 97-59-6; EC No: 202-592-8; MDL No: MFCD00005260; RTECS: YT1600000 Solid; Molecular Formula: C4H6N4O3 ; MW: 158.12 Melting Point: 230° (decomposes)

  • RTECS: YT1600000
  • Solid


Formula: C₄H₆N₄O₃
Melting Pt: 230 °C (dec.)
MDL Number: MFCD00005260
CAS Number: 97-59-6
EINECS: 202-592-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty