Copper(II) carbonate basic
Supplier: BEANTOWN CHEMICAL - BTC
142190-2KG
142190-500G
BT142190-2KGEA
146
USD
BT142190-2KG
BT142190-500G
Copper(II) carbonate basic
Copper(II) carbonate basic
CAS: 12069-69-1; EC No: 235-113-6; MDL No: MFCD00010976; RTECS: GL6910000
Powder; Linear Formula: CuCO3·Cu(OH)2; MW: 221.11
Melting Point: 200° (decomposes)
- RTECS: GL6910000
- Powder
Formula:
CuCO₃·Cu(OH)₂ MW: 221.12 g/mol Melting Pt: 200 (dec.) |
MDL Number:
MFCD00010976 CAS Number: 12069-69-1 EINECS: 235-113-6 UN: 3288 ADR: 6.1 III Merck Index: 13,02658 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...