2-Amino-6-chloropurine riboside ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

143110-1G 143110-5G
BT143110-1GEA 121 USD
BT143110-1G BT143110-5G
2-Amino-6-chloropurine riboside ≥95%
2-Amino-6-chloropurine riboside

CAS: 2004-07-1; EC No: 217-905-3; MDL No: MFCD00005735 Crystalline/Solid; Molecular Formula: C10H12ClN5O4; MW: 301.69 Melting Point: 165-167° (decomposes)


  • Crystalline/Solid

Formula: C₁₀H₁₂ClN₅O₄
MW: 301.69 g/mol
Melting Pt: 165…167 °C (dec.)
MDL Number: MFCD00005735
CAS Number: 2004-07-1
EINECS: 217-905-3

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR