Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-O-Palmitoyl-L-ascorbic acid ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

143650-100G 143650-25G 143650-500G
BT143650-100GEA 100.7 USD InStock
BT143650-100G BT143650-25G BT143650-500G
6-O-Palmitoyl-L-ascorbic acid ≥99%
6-O-Palmitoyl-L-ascorbic acid
CAS: 137-66-6; EC No: 205-305-4; MDL No: MFCD00005377 Crystalline; Molecular Formula: C22H38O7; MW: 414.52

  • Crystalline


Formula: C₂₂H₃₈O₇
MW: 414.54 g/mol
MDL Number: MFCD00005377
CAS Number: 137-66-6
EINECS: 205-305-4
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty