Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium bromide, anhydrous ≥99.5%

Supplier: BEANTOWN CHEMICAL - BTC

145425-10G 145425-250G 145425-50G
BT145425-10GEA 36.19 USD InStock
BT145425-10G BT145425-250G BT145425-50G
Calcium bromide, anhydrous ≥99.5%
Calcium bromide
CAS: 7789-41-5; EC No: 232-164-6; MDL No: MFCD00010902; RTECS: EV9328000 -80 Mesh Powder; Linear Formula: CaBr2; MW: 199.89 Density (g/mL): 3.353 Hygroscopic

  • RTECS: EV9328000
  • Hygroscopic
  • -80 Mesh Powder


Formula: Br₂Ca
Density: 3.353 g/mL
MDL Number: MFCD00010902
CAS Number: 7789-41-5
EINECS: 232-164-6
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty