Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2,4-dinitrobenzenesulphonate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

217820-25G 217820-5G
BT217820-25GEA 72 USD InStock
BT217820-25G BT217820-5G
Sodium-2,4-dinitrobenzenesulphonate ≥97%
Sodium-2,4-dinitrobenzenesulphonate
CAS: 885-62-1; EC No: 212-943-7; MDL No: MFCD00007471 Powder; Molecular Formula: C6H3N2NaO7S; MW: 270.15

  • Powder


Formula: C₆H₃N₂NaO₇S
MDL Number: MFCD00007471
CAS Number: 885-62-1
EINECS: 212-943-7
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty