Molybdenum ≥99.95% (trace metals basis), rods Ø 3.17 mm

Supplier: BEANTOWN CHEMICAL - BTC

218580-50CM 218580-5X50CM
BT218580-50CMEA 50 USD
BT218580-50CM BT218580-5X50C
Molybdenum ≥99.95% (trace metals basis), rods Ø 3.17 mm
Molybdenum

CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465 Solid; Molecular Formula: Mo; MW: 95.94 Melting Point: 2617°; Boiling Point: 4612° Density (g/mL): 10.3


  • Solid

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.3 g/mL
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR