Bromocresol purple
Supplier: BEANTOWN CHEMICAL - BTC
220260-10G
220260-50G
BT220260-10GEA
34.63
USD
BT220260-10G
BT220260-50G
Bromocresol purple
Bromocresol purple
CAS: 115-40-2; EC No: 204-087-8; MDL No: MFCD00011681
Solid; Molecular Formula: C21H16Br2O5S; MW: 540.22
Melting Point: 240° (decomposes)
- Solid
Formula:
C₂₁H₁₆Br₂O₅S MW: 540.23 g/mol Melting Pt: 242 (dec.) |
MDL Number:
MFCD00011681 CAS Number: 115-40-2 EINECS: 204-087-8 Merck Index: 13,01371 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...