Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3,5-dichloroisonicotinic acid ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

222690-1G 222690-250MG 222690-5G
BT222690-1GEA 88 USD InStock
BT222690-1G BT222690-250MG BT222690-5G
2-Bromo-3,5-dichloroisonicotinic acid ≥95%
2-Bromo-3,5-dichloroisonicotinic acid
CAS: 343781-56-6 Solid; Molecular Formula: C6H2BrCl2NO2 ; MW: 270.90

  • Solid


Formula: C₆H₂BrCl₂NO₂
MDL Number: MFCD03411637
CAS Number: 343781-56-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty