Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MPC-11]

Supplier: Biolegend
400311 400312 400313 400314
400311-BLEA 60.3 USD InStock
400311-BL 400312-BL 400313-BL 400314-BL
Anti-IgG2b, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MPC-11]
Antibodies
PE Mouse IgG2b, κ Isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests

Each lot of this mouse IgG2b, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG2b, k Isotype Ctrl
Clonality: Monoclonal
Clone: MPC-11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty