Anti-IgM µ Goat Polyclonal Antibody (CF™ 647)

Supplier: Biotium
20346-250UL 20346-50UL
89427-206EA 193.3 USD
89427-206 89427-208
Anti-IgM µ Goat Polyclonal Antibody (CF™ 647)
Antibodies
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: CF™ 647
Epitope: µ-chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR