Anti-IgG Donkey Polyclonal Antibody (CF™ 543)

Supplier: Biotium
20305 20305-1
89411-042EA 260.49 USD
89411-042 89411-490
Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR