Anti-IgG Donkey Polyclonal Antibody (CF™ 543)

Supplier: Biotium
20308 20308-1
89411-048EA 322.57 USD
89411-048 89411-496
Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Order Now

SPECIFICATIONS

Antigen Symbol IgG
Environmentally Preferable
Antigen Name Immunoglobulin G
Antibody Type Secondary
Clonality Polyclonal
Conjugation CF® 543
Reactivity Rabbit
Host Donkey
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Epitope Heavy and Light chain
Cross Adsorption Minimal Cross-Reactivity: Bv, Ch, Gt, GP, Hs, Hu, Ms, Sh, SHm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR