Anti-IgG Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium
20328 20328-1
89411-088EA 338.03 USD
89411-088 89411-536
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR