A0717-25G
A0717-5G
TCA0717-25GEA
192.13
USD
InStock
TCA0717-25G
TCA0717-5G
1-Adamantanecarbonyl chloride ≥97.0% (by GC, titration analysis)
1-Adamantanecarbonyl chloride
CAS Number: 2094-72-6
MDL Number: MFCD00074724
Molecular Formula: C11H15ClO
Molecular Weight: 198.69
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 53
Flash Point (°C): 110