Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Propylthio)propene ≥97.0% (by GC)

Supplier: Decon Laboratories Ltd

A1061-25ML A1061-5ML
TCA1061-25MLEA 235 USD InStock
TCA1061-25ML TCA1061-5ML
3-(Propylthio)propene ≥97.0% (by GC)
3-(Propylthio)propene
CAS Number: 27817-67-0
MDL Number: MFCD00015220
Molecular Formula: C6H12S
Molecular Weight: 116.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 0.87


Formula: C₆H₁₂S
Boiling Pt: 140 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00015220
CAS Number: 27817-67-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cea40cf6635bd96f8d4442c4c9d896125a767602&vid=cea40cf6635bd96f8d4442c4c9d896125a767602&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty