Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltrimethylsilane ≥96.0%

Supplier: Decon Laboratories Ltd

B1856-25ML B1856-5ML
TCB1856-25MLEA 146 USD InStock
TCB1856-25ML TCB1856-5ML
Benzyltrimethylsilane ≥96.0%
Benzyltrimethylsilane
CAS Number: 770-09-2
MDL Number: MFCD00039796
Molecular Formula: C10H16Si
Molecular Weight: 164.32
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Flash Point (°C): 62
Specific Gravity (20/20): 0.87

Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: alpha-Trimethylsilyltoluene


Formula: C₁₀H₁₆Si
Boiling Pt: 191 °C
Density: 0.87 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00039796
CAS Number: 770-09-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=252101a5cb1c4c5848f1ecbd0f58207621f69b7b&vid=252101a5cb1c4c5848f1ecbd0f58207621f69b7b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty