Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromoacetamide ≥98.0% (by GC, total nitrogen)

Supplier: Decon Laboratories Ltd

B2852-25G B2852-5G
TCB2852-25GEA 134 USD InStock
TCB2852-25G TCB2852-5G
Bromoacetamide ≥98.0% (by GC, total nitrogen)
Bromoacetamide
CAS Number: 683-57-8
MDL Number: MFCD00008025
Molecular Formula: C2H4BrNO
Molecular Weight: 137.96
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 89


Formula: BrCH₂CONH₂
Melting Pt: 89 °C
MDL Number: MFCD00008025
CAS Number: 683-57-8
UN: 2923
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36a25515753af0b3bd177bc81ea26edde3e1f7cf&vid=36a25515753af0b3bd177bc81ea26edde3e1f7cf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty