Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Butanedithiol ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

B2888-25G B2888-5G
TCB2888-25GEA 108 USD InStock
TCB2888-25G TCB2888-5G
2,3-Butanedithiol ≥98.0% (by GC)
2,3-Butanedithiol
CAS Number: 4532-64-3
MDL Number: MFCD00010024
Molecular Formula: C4H10S2
Molecular Weight: 122.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Flash Point (°C): 52
Specific Gravity (20/20): 1.03

Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Monomers
Other Product Names / Synonyms: 2,3-Dimercaptobutane


Formula: C₄H₁₀S₂
Boiling Pt: 87 °C
Density: 1.03 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00010024
CAS Number: 4532-64-3
UN: 3336
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1e97ee36b7f54be3ea2e19fde33707b94ca9853&vid=f1e97ee36b7f54be3ea2e19fde33707b94ca9853&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty