Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butylphenyl isocyanate ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

B3205-1G B3205-5G
TCB3205-1GEA 30 USD InStock
TCB3205-1G TCB3205-5G
4-Butylphenyl isocyanate ≥98.0% (by GC)
4-Butylphenyl isocyanate
CAS Number: 69342-47-8
MDL Number: MFCD00013882
Molecular Formula: C11H13NO
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 108
Specific Gravity (20/20): 1.00

Other Product Names / Synonyms: Isocyanic Acid 4-Butylphenyl Ester


Formula: C₁₁H₁₃NO
Boiling Pt: 170 °C
Density: 1.00 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00013882
CAS Number: 69342-47-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9436f273e80b4a3ad5d85d4a9d5321362c0fb1b&vid=c9436f273e80b4a3ad5d85d4a9d5321362c0fb1b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty