Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorobenzenesulphonyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd

B3698-25G B3698-5G
TCB3698-25GEA 340 USD InStock
TCB3698-25G TCB3698-5G
4-Bromo-2-fluorobenzenesulphonyl chloride ≥98.0%
4-Bromo-2-fluorobenzenesulphonyl chloride
CAS Number: 216159-03-4
MDL Number: MFCD01090993
Molecular Formula: C6H3BrClFO2S
Molecular Weight: 273.50
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 66


Formula: C₆H₃BrClFO₂S
Melting Pt: 66 °C
MDL Number: MFCD01090993
CAS Number: 216159-03-4
UN: 3261
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1908f9a3a4535bea807de9a827ee68936091451&vid=a1908f9a3a4535bea807de9a827ee68936091451&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty