D0117-25G
D0117-5G
TCD0117-25GEA
483.3
USD
InStock
TCD0117-25G
TCD0117-5G
2,3-Diaminopyridine ≥98.0%
2,3-Diaminopyridine
CAS Number: 452-58-4
MDL Number: MFCD00006319
Molecular Formula: C5H7N3
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 114
Flash Point (°C): 205