Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluorophenol ≥98.0% (by GC)

Supplier: Decon Laboratories Ltd

D1832-25G D1832-5G
TCD1832-25GEA 112 USD InStock
TCD1832-25G TCD1832-5G
2,4-Difluorophenol ≥98.0% (by GC)
2,4-Difluorophenol
CAS Number: 367-27-1
MDL Number: MFCD00009715
Molecular Formula: C6H4F2O
Molecular Weight: 130.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 52
Flash Point (°C): 56
Freezing point (°C): 22


Formula: C₆H₄F₂O
Boiling Pt: 52 °C
Flash Pt: 56 °C
MDL Number: MFCD00009715
CAS Number: 367-27-1
UN: 2921
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e491e8049f04f21e6ed2d99e68afdb27ecaf0ada&vid=e491e8049f04f21e6ed2d99e68afdb27ecaf0ada&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty