D2953-25G
D2953-5G
TCD2953-25GEA
100
USD
InStock
TCD2953-25G
TCD2953-5G
2,5-Dichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
2,5-Dichlorobenzoyl chloride
CAS Number: 2905-61-5
MDL Number: MFCD00035821
Molecular Formula: C7H3Cl3O
Molecular Weight: 209.45
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 95
Freezing point (°C): 23