Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2S,3S)-diylbis(methylammonium)tetrafluoroborate [=(S,S)-TaDiAS-2nd] ≥95.0%

Supplier: Decon Laboratories Ltd

D3476-1G D3476-200MG
TCD3476-1GEA 660 USD InStock
TCD3476-1G TCD3476-200MG
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2S,3S)-diylbis(methylammonium)tetrafluoroborate [=(S,S)-TaDiAS-2nd] ≥95.0%
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2S,3S)-diylbis(methylammonium)tetrafluoroborate [=(S,S)-TaDiAS-2nd]
Molecular Weight: 1000.82
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Molecular Formula: C58H70B2F8N2O2

Other Product Names / Synonyms: (S,S)-TaDiAS-2nd


Formula: C₅₈H₇₀B₂F₈N₂O₂
CAS Number: 2135524-59-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79fc7a495de092df53d329b3c6dd2fcf44ecdcfc&vid=79fc7a495de092df53d329b3c6dd2fcf44ecdcfc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty