Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Indanol ≥99.0% (by GC)

Supplier: Decon Laboratories Ltd

H0251-100G H0251-25G
TCH0251-100GEA 268.63 USD InStock
TCH0251-100G TCH0251-25G
5-Indanol ≥99.0% (by GC)
5-Indanol
CAS Number: 1470-94-6
MDL Number: MFCD00003802
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 255
Melting point (°C): 53
Flash Point (°C): 130

Other Product Names / Synonyms: 5-Hydroxyhydrindene, 5-Indanol


Formula: C₉H₁₀O
Boiling Pt: 255 °C
Melting Pt: 53 °C
Flash Pt: 130 °C
MDL Number: MFCD00003802
CAS Number: 1470-94-6
UN: 2811
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18e5e0d5477e7b5061d58a457a0cfc10c455ee5f&vid=18e5e0d5477e7b5061d58a457a0cfc10c455ee5f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty