Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodobenzenesulfonyl chloride ≥93.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

I0051-25G I0051-5G
TCI0051-25GEA 154 USD InStock
TCI0051-25G TCI0051-5G
4-Iodobenzenesulfonyl chloride ≥93.0% (by titrimetric analysis)
4-Iodobenzenesulphonyl chloride
CAS Number: 98-61-3
MDL Number: MFCD00007441
Molecular Formula: C6H4ClIO2S
Molecular Weight: 302.51
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Melting point (°C): 80

Other Product Names / Synonyms: 4-Iodophenylsulfonyl Chloride, Pipsyl Chloride


Formula: C₆H₄ClIO₂S
Melting Pt: 80 °C
MDL Number: MFCD00007441
CAS Number: 98-61-3
UN: 3261
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf138280e398ca9cd00159705dbceaf4e0680673&vid=cf138280e398ca9cd00159705dbceaf4e0680673&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >93.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty