I0558-1G
I0558-5G
TCI0558-1GEA
26
USD
InStock
TCI0558-1G
TCI0558-5G
Ethyl isothiocyanatoacetate ≥98.0% (by GC)
Ethyl isothiocyanatoacetate
CAS Number: 24066-82-8
MDL Number: MFCD00060677
Molecular Formula: C5H7NO2S
Molecular Weight: 145.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 109
Specific Gravity (20/20): 1.16
Other Product Names / Synonyms: Isothiocyanatoacetic Acid Ethyl Ester, Ethoxycarbonylmethyl Isothiocyanate