R0007-1G
R0007-5G
TCR0007-1GEA
92.5
USD
InStock
TCR0007-1G
TCR0007-5G
Reserpine ≥98.0% (by HPLC, titration analysis)
Reserpine
CAS Number: 50-55-5
MDL Number: MFCD00005091
Molecular Formula: C33H40N2O9
Molecular Weight: 608.69
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow
Specific rotation [a]20/D: 177 deg (C=1, DMF)